$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT HUMAN CASP P AB202201-100UG
ABCA_AB202201-100U
:
Abcam
New Product
RECOMBINANT HUMAN CASP P AB202201-100UG
ABCA_AB202201-100U
:
Abcam
:
AB202201-100UG
New Product