α-Cyclodextrin non-reducing cyclic saccharide
Product Details & Documents
Non-reducing cyclic saccharide consisting of six α-1,4-linked D-glucopyranosyl units produced by the action of cyclodextrin glucosyltransferase (CGTase, EC 2.4.1.19) on hydrolyzed starch. Useful for the solubilization of nonpolar macromolecules such as fatty acids, lipids, and cholesterol for use in cell culture applications. Cyclodextrins are low molecular weight organic molecules which form water-soluble complexes with various compounds.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:10016-20-3
- Molecular Weight:972.86 g/mol
Specifications
Specification Test Results
- Constituents::100% a-Cyclodextrin hydrate
Specifications
- Size:25 g
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



