$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT HUMAN AMPK ALP AB185258-5UG
ABCA_AB185258-5UG
:
Abcam
New Product
RECOMBINANT HUMAN AMPK ALP AB185258-5UG
ABCA_AB185258-5UG
:
Abcam
:
AB185258-5UG
New Product