Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HUMAN AMPK AL AB177266-20UG
Catalog # ABCA_AB177266-20UG
Supplier:
Abcam
New Product
Expand image
RECOMBINANT HUMAN AMPK AL AB177266-20UG
Catalog # ABCA_AB177266-20UG
Supplier:
Abcam
Supplier Number:
AB177266-20UG
New Product