RIPA Buffer, Abcam
RIPA Buffer, 500 ml
Catalog # ABCA_AB288006-500M
New Product
Supplier: Abcam
Supplier #: AB288006-500ML
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.
- Constituents: 1% Alcohol Ethoxylate, 0.877% Sodium chloride, 0.788% Tris HCl, 0.5% Sodium deoxycholate, 0.1% Sodium lauryl sulfate
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:RIPA Buffer
- Size:500 ml
- Cat. No.:ABCA_AB288006-500M
- Storage Temperature:Store at -20°C.



