Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, 5 ml
Catalog # ABCA_AB193971-5ML
New Product
Supplier:   Abcam
Supplier #:  AB193971-5ML
Extraction Enhancer Buffer 50X, Abcam
Catalog # ABCA_AB193971-5ML
Supplier:  Abcam
New Product
CAS Number:  
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, 5 ml

CAS Number:  
Price
$52.70
Log in for availability
Each (5ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Abcam's Extraction Enhancer Buffer 50X is formulated for the preparation of cell or tissues extracts for use in immunoassays.

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Extraction Enhancer Buffer 50X
  • Size:
    5 ml
  • Cat. No.:
    ABCA_AB193971-5ML
  • Storage Temperature:
    Store at +4°C.

Specifications