CELL LINE CACO-2 CACO2 ENTEROCYTE FRZN
Catalog # MSPP-HTB-37
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL LINE CACO-2 CACO2 ENTEROCYTE FRZN
Catalog # MSPP-HTB-37
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: HTB-37
Frequently Bought Together
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Western Blotting Kit, Azure Biosystems
Supplier: Azure Biosystems
PI MARKER 9.50 200UL
Supplier: PROTEIN SIMPLE MS
Endo-Wash Buffer, Zymo Research
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Peacock™ Prestained Protein Markers, Biotium
Supplier: Biotium
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
PI MARKER ICE280/ICE3 6.61 200UL
Supplier: PROTEIN SIMPLE MS
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
Zyppy Wash Buffer, Zymo Research
Supplier: Zymo Research
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
BUFFER LDS SAMPLE LOADING (4X) 10ML
Supplier: G-Biosciences
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Perfect Protein™ Markers, MilliporeSigma
Supplier: MilliporeSigma
AzureSpectra Fluorescent Western Blotting Kits, Azure Biosystems
Supplier: Azure Biosystems