Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
(4-Bromo-2,3-dichlorophenyl)(phenyl)methanol ≥95%

(4-Bromo-2,3-dichlorophenyl)(phenyl)methanol ≥95%

Glass bottle, 1 g
Catalog # 77934-199
New Product
Supplier:   AOBChem USA
Supplier #:  567813-1G
(4-Bromo-2,3-dichlorophenyl)(phenyl)methanol ≥95%
Catalog # 77934-199
Supplier:  AOBChem USA
New Product
CAS Number:  
(4-Bromo-2,3-dichlorophenyl)(phenyl)methanol ≥95%

Glass bottle, 1 g

CAS Number:  
Price
$403.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: For research use only

  • Formula:C₁₃H₉BrCl₂O
  • Molecular Weight:332.02
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.