Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

D-Glucamine ≥97% (by titrimetric analysis)

5 g, Plastic bottle
Catalog # CH04463-5G
New Product
 :  04463-5G
D-Glucamine ≥97% (by titrimetric analysis)
Catalog # CH04463-5G
New Product
CAS Number:  488-43-7
D-Glucamine ≥97% (by titrimetric analysis)
D-Glucamine ≥97% (by titrimetric analysis)

5 g, Plastic bottle

CAS Number:  488-43-7
Price
$213.62
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

D-Glucamine is a versatile amino sugar that plays a crucial role in various industrial and research applications. Known for its unique structure, D-Glucamine is recognized for its ability to function as a chelating agent, making it particularly valuable in the formulation of pharmaceuticals and dietary supplements. Its properties allow it to enhance the solubility and bioavailability of active ingredients, which is essential for effective drug delivery systems. Additionally, D-Glucamine is utilized in the food industry as a sweetener and stabilizer, providing a healthier alternative to traditional sugars while maintaining desirable taste profiles.

Researchers appreciate D-Glucamine for its potential in biochemistry and molecular biology, where it serves as a building block for synthesizing various compounds. Its compatibility with biological systems makes it an ideal candidate for developing innovative therapeutic agents. With its multifunctional capabilities and broad applicability, D-Glucamine stands out as a valuable compound for professionals seeking to enhance product formulations and explore new avenues in research.

Store at 0 to 8 °C.

Synonyms:  1-Amino-1-deoxy-D-glucitol, D-Glucamine, 1-Amino-1-deoxy-D-sorbitol, (-)-1-Amino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-Aminohexane-1,2,3,4,5-pentaol
  • CAS Number:488-43-7
  • Formula:C₆H₁₅NO₅
  • MDL Number:MFCD00077776
  • Molecular Weight:181.2 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.