Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Basic fuchsin ≥95%

Basic fuchsin ≥95%

Plastic bottle, 5 kg
Catalog # CH01710-5KG
New Product
Supplier #:  01710-5KG
Basic fuchsin ≥95%
Catalog # CH01710-5KG
New Product
CAS Number:  632-99-5
Basic fuchsin ≥95%
Basic fuchsin ≥95%

Plastic bottle, 5 kg

CAS Number:  632-99-5
Price
$1,162.65
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Basic fuchsin is a vibrant synthetic dye widely utilized in various industries, particularly in histology and microbiology for staining applications. Known for its strong affinity for biological tissues, it is commonly employed in the Gram staining process, allowing researchers to differentiate between Gram-positive and Gram-negative bacteria effectively. Its unique properties enable clear visualization of cellular structures under a microscope, making it an invaluable tool in laboratory settings. Additionally, Basic fuchsin serves as a pH indicator and is used in textile dyeing, providing bright coloration to fabrics while maintaining excellent lightfastness.

The compound's versatility extends to its use in the production of colored plastics and as a marker in various chemical reactions. Its stability and ease of application make it a preferred choice among professionals seeking reliable and effective staining solutions. With its broad range of applications, Basic fuchsin stands out for its ability to enhance the clarity of microscopic observations and its role in quality control processes across multiple industries.

Shipping temperature: Store at RT

Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀N₃Cl
  • MDL Number:MFCD00012569
  • Molecular Weight:337.86 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.