Orders LinkContactUsLinkComponent
a-Cyclodextrin USP
Plastic bottle, 25 g
  CH01963-25G
New Product
 :  01963-25G
a-Cyclodextrin USP
  CH01963-25G
New Product
 :  10016-20-3
a-Cyclodextrin USP
a-Cyclodextrin USP
Plastic bottle, 25 g
 :  10016-20-3
$39.59
Each(1/Itm/Bx)
a-Cyclodextrin is a cyclic oligosaccharide composed of seven glucose units, known for its unique ability to form inclusion complexes with various guest molecules. This property makes it an invaluable compound in the pharmaceutical, food, and cosmetic industries. Its ability to enhance the solubility and stability of poorly soluble drugs allows for improved bioavailability, making it a popular choice in drug formulation. Additionally, a-Cyclodextrin is utilized in food applications to encapsulate flavors and aromas, thereby enhancing product quality and shelf life. In cosmetics, it serves as a stabilizer and carrier for active ingredients, ensuring effective delivery and prolonged efficacy.
Moreover, a-Cyclodextrin's non-toxic and biodegradable nature aligns with the growing demand for sustainable and safe ingredients in various applications. Its versatility extends to research, where it is employed in analytical chemistry for separation and purification processes. With its broad range of applications and benefits, a-Cyclodextrin stands out as a crucial ingredient for professionals seeking to innovate and improve product formulations across multiple industries.
Store at RT.
  •  :10016-20-3
  •  :C₃₆H₆₀O₃₀
  •  :MFCD00078207
  •  :972.9 g/mol
  •  :Room Temperature
  • Pack type:
    Plastic bottle
  • Size:
    25 g