Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L-Alanine 98.5-101.5% (dried basis) USP

L-Alanine 98.5-101.5% (dried basis) USP

Plastic bottle, 100 g
Catalog # CH00019-100G
New Product
Supplier #:  00019-100G
L-Alanine 98.5-101.5% (dried basis) USP
Catalog # CH00019-100G
New Product
CAS Number:  56-41-7
L-Alanine 98.5-101.5% (dried basis) USP
L-Alanine 98.5-101.5% (dried basis) USP

Plastic bottle, 100 g

CAS Number:  56-41-7
Price
$30.33
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

L-Alanine is a non-essential amino acid that plays a crucial role in protein synthesis and metabolic processes. It is recognized for its ability to support muscle metabolism and enhance energy production, making it a valuable compound in sports nutrition and dietary supplements. L-Alanine is often utilized in the formulation of protein powders and energy drinks, where it aids in the maintenance of muscle mass and recovery post-exercise. Additionally, it serves as a precursor for glucose production, which can be particularly beneficial during prolonged physical activity or fasting states.

In the pharmaceutical and research sectors, L-Alanine is employed in various applications, including the synthesis of pharmaceuticals and as a building block in peptide synthesis. Its unique properties, such as its ability to stabilize protein structures and its low toxicity, make it an attractive option for researchers and formulators. With its versatile applications in nutrition, pharmaceuticals, and biochemistry, L-Alanine stands out as a compound that supports both health and performance.

Store at room temperature.

Synonyms:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS Number:56-41-7
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00064410
  • Molecular Weight:89.09 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.