Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lincomycin hydrochloride

Lincomycin hydrochloride

Plastic bottle, 5 g
Catalog # CH00272-5G
New Product
Supplier #:  00272-5G
Lincomycin hydrochloride
Catalog # CH00272-5G
New Product
CAS Number:  859-18-7
Lincomycin hydrochloride
Lincomycin hydrochloride

Plastic bottle, 5 g

CAS Number:  859-18-7
Price
$111.76
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Lincomycin hydrochloride is a potent antibiotic widely utilized in the treatment of various bacterial infections, particularly those caused by anaerobic bacteria and certain protozoa. This compound is especially valuable in clinical settings for its effectiveness against infections in patients who are allergic to penicillin. Lincomycin hydrochloride works by inhibiting bacterial protein synthesis, making it a critical option for treating serious infections such as osteomyelitis, pneumonia, and skin infections. Its unique mechanism of action allows it to penetrate well into tissues, providing effective treatment in hard-to-reach areas of the body.

In addition to its therapeutic applications, Lincomycin hydrochloride is also used in veterinary medicine, particularly for treating infections in livestock. Its ability to combat resistant strains of bacteria makes it an essential tool in both human and animal healthcare. Researchers continue to explore its potential in combination therapies, enhancing its efficacy against multi-drug resistant pathogens. With its established safety profile and versatility, Lincomycin hydrochloride remains a cornerstone in antibiotic therapy, offering reliable solutions for healthcare professionals.

Store at 0 to 8 °C.

  • CAS Number:859-18-7
  • Formula:C₁₈H₃₄N₂O₆S·HCl
  • MDL Number:MFCD00083647
  • Molecular Weight:443 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.