ContactUsLinkComponent
COVERGLASS PREMIUM #1 24X55 1OZ CS10
Premium Coverglass #1
Catalog # 77908-134
Supplier:  StatLab
New Product
Premium Coverglass #1
Catalog # 77908-134
Supplier:  StatLab
Supplier Number:  SL102455
New Product
Specifications
  • Description:
    Premium coverglass #1
  • L×W:
    24×55 mm
  • Environmentally Preferable:
Specifications
About this item
Premium coverglass specially designed and manufactured to work with automatic coverslippers.