ContactUsLinkComponent
MAST CELL CONTROLS PK25
Mast Cell IHC Control Slides
Catalog # 77907-930
Supplier:  StatLab
New Product
undefined
Mast Cell IHC Control Slides
Catalog # 77907-930
Supplier:  StatLab
Supplier Number:  CSM0125P
New Product
Specifications
  • Description:
    Mast cell IHC control slides
  • Environmentally Preferable:
Specifications
About this item
Mast Cell IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included Tissue: known positive Special stain or antibody: Mast Cell
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.