Orders LinkContactUsLinkComponent
CellSafe+ Cassette Biopsy Capsule
CellSafe+ Cassette Biopsy Capsule
Cellsafe+ cassette biopsy capsule, White
Catalog # 77908-040
New Product
Supplier:   StatLab
Supplier #:  EBE-0201-02A
CellSafe+ Cassette Biopsy Capsule
Catalog # 77908-040
Supplier:  StatLab
New Product
CellSafe+ Cassette Biopsy Capsule
Cellsafe+ cassette biopsy capsule, White
Price
$121.13
Log in for availability
Case of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cellsafe+ is an easy-to-use alternative to the more traditional and time-consuming handling techniques such as tissue wrap and foam pads.
  • CellSafe+ is contained in a standard cassette for normal processing Validated for use with standard cassettes
The CellSafe+ consists of two hinged interlocking frames with integral mesh. This extra fine x-ray transparent mesh ensures a high level of specimen security, zero tissue damage, and low reagent carry over. The capsule is closed in one direction to fully encapsulate the tissue. Prior to embedding, tissue can be removed with greater ease and security, compared with conventional foam pads or paper wrap.
The CellSafe+ biopsy capsule is a patented innovation by CellPath, a StatLab company.
Specifications
  • Color:
    White
  • Description:
    Cellsafe+ cassette biopsy capsule