Orders LinkContactUsLinkComponent
PathFlow Supa Mega Slim Cassettes
PathFlow Supa Mega Slim Cassettes
PathFlow supa mega slim cassettes, Red
Catalog # 77908-024
New Product
Supplier:   StatLab
Supplier #:  EAN-0110-02A
PathFlow Supa Mega Slim Cassettes
Catalog # 77908-024
Supplier:  StatLab
New Product
PathFlow Supa Mega Slim Cassettes
PathFlow supa mega slim cassettes, Red
Price
$266.21
Log in for availability
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
  • Casting can be done in 20 minutes at −8 °C
  • Large color range available
  • Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
Specifications
  • Color:
    Red
  • Description:
    PathFlow supa mega slim cassettes