ContactUsLinkComponent
CLAMP PATHFLOW UNIVERSAL SUPA MEGA
PathFlow Universal Supa Mega Cassette Clamp
Catalog # 77908-068
Supplier:  StatLab
New Product
PathFlow Universal Supa Mega Cassette Clamp
Catalog # 77908-068
Supplier:  StatLab
Supplier Number:  JFB-0100-00A
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PathFlow universal supa mega cassette clamp
  • Environmentally Preferable:
Specifications
About this item
The Supa Mega cassette clamp is used for the microtomy of large tissue blocks.
  • Quick and easy to fit to the Leica and Epredia microtomes
  • Incorporated spirit level ensures correct alignment when attached to the microtome
  • Secure clamping of the cassette
  • Easy to use quick release function
  • Innovative design avoids collision between the clamp and knife block
  • Highly durable
Incorporating an intuitive quick-release system, the clamp can securely hold any tissue processing cassette from the Supa Mega range (Supa Mega Slim Cassette with spacer). Unlike other large tissue block clamps, the Supa Mega clamp’s unique design allows sectioning all the way through the Supa Mega block without collision between the clamp and knife block, which opens up the possibility of using Supa Mega Slim and Supa Mega Mothership base molds. The design also prevents the possibility of the cassette being attached in the wrong orientation. Easily fits Leica Biosystems' HistoCore, RM2100 and RM2200, and Epredia's Microm HH series of microtomes.