PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 60×45×8 mm
Catalog # 77908-060
New Product
Supplier: StatLab
Supplier #: GBC-6014-05B
Jump to:
Product Details & Documents
PathFlow stainless steel Supa Mega base molds.
- Reusable Stops wax leakage Stackable design for efficient storage Stainless steel for a lifetime of use
Specifications
- Description:PathFlow Supa Mega base mold
- Dimensions:60×45×8 mm