Orders LinkContactUsLinkComponent
PathFlow Supa Mega Base Mold
PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 60×45×15 mm
Catalog # 77908-058
New Product
Supplier:   StatLab
Supplier #:  GBC-6014-05A
PathFlow Supa Mega Base Mold
Catalog # 77908-058
Supplier:  StatLab
New Product
PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 60×45×15 mm
Price
$368.49
Log in for availability
Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PathFlow stainless steel Supa Mega base molds.
  • Reusable Stops wax leakage Stackable design for efficient storage Stainless steel for a lifetime of use
Specifications
  • Description:
    PathFlow Supa Mega base mold
  • Dimensions:
    60×45×15 mm