Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

γ-Cyclodextrine ≥98%

Bottle, 500 g
Catalog # ALC103968-500G
Irritant
New Product
Supplier:   Aladdin Scientific
 :  C103968-500G
γ-Cyclodextrine ≥98%
Catalog # ALC103968-500G
Supplier:  Aladdin Scientific
New Product
CAS Number:  17465-86-0
γ-Cyclodextrine ≥98%

Bottle, 500 g

CAS Number:  17465-86-0
Price
$217.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:241-482-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.