Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
β-Cyclodextrine ≥98%

β-Cyclodextrine ≥98%

Plastic bottle, 500 g
Catalog # ALC104384-500G
Irritant
New Product
Supplier:   Aladdin Scientific
Supplier #:  C104384-500G
β-Cyclodextrine ≥98%
Catalog # ALC104384-500G
Supplier:  Aladdin Scientific
New Product
CAS Number:  7585-39-9
β-Cyclodextrine ≥98%

Plastic bottle, 500 g

CAS Number:  7585-39-9
Price
$39.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Useful for forming water-soluble complexes for fluorescent labeling of proteins. Use to solubilize non-polar compounds such as fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomersβ-Cyclodextrin is used with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins. It is an active ingredient of household odor eliminator.

It is also used in personal care products like toothpastes, skin creams and dusting powders. It finds applications in the cosmetic industry for products like detergents and perfumes for the controlled release of fragrances. Further, it is used to produce HPLC columns allowing chiral enantiomers separation. In addition to this, it is used to decrease the level of cholesterol in milk fat.

Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.