Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Cleaning Validation Coupons

Cleaning validation coupon, Steel, hard chrome plated, 152.4×152.4 mm
Catalog # 77887-708
New Product
Supplier:   GLOBEPHARMA
 :  8001900
Cleaning Validation Coupons
Catalog # 77887-708
Supplier:  GLOBEPHARMA
New Product
Cleaning Validation Coupons
Cleaning Validation Coupons

Cleaning validation coupon, Steel, hard chrome plated, 152.4×152.4 mm

Price
$309.98
Log in for availability
Each
Product Details & Documents

Cleaning validation coupons are used in the laboratory to validate a proposed swabbing method.

  • All popular sizes available

  • Wide variety of materials and finishes

GlobePharma offers cleaning validation coupons made from a wide range of materials, including:

316 and 304 stainless steel
Aluminum
Brass and bronze
Various plastics and rubbers

These coupons are available in standard sizes of  0.32×5.1×5.1 cm (⅛×2×2”) and 0.32×10.2×10.2 cm (⅛×4×4”).

Specifications
  • Description:
    Cleaning validation coupon
  • Material:
    Steel, hard chrome plated
  • Size:
    152.4×152.4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.