Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GAUGE DIG PRS 0.5% 1/4NPT SS 5PSI BAT
Catalog # MFLX33000-24
Supplier:
KODIAK CONTROLS
New Product
Expand image
GAUGE DIG PRS 0.5% 1/4NPT SS 5PSI BAT
Catalog # MFLX33000-24
Supplier:
KODIAK CONTROLS
Supplier Number:
MG-5-A-9V-R
New Product