Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE E18 MOUSE SUBSTANTIA NIGRA PAIR-1
Catalog # MSPP-C57ESN
Supplier:
BRAINBITS LLC MS
Expand image
TISSUE E18 MOUSE SUBSTANTIA NIGRA PAIR-1
Catalog # MSPP-C57ESN
Supplier:
BRAINBITS LLC MS
Supplier Number:
C57ESN