$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER TAGMENTATION 10000UL
MSPP-C010190-4
:
Diagenode
BUFFER TAGMENTATION 10000UL
MSPP-C010190-4
:
Diagenode
:
C01019043-10000