Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Fluorometer

VWR® Fluorometer

VWR® Fluorometer, 100 - 240 VAC, Excitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nm
  VWRI210-000157
VWR® Fluorometer
  VWRI210-000157
VWR® Fluorometer
VWR® Fluorometer

VWR® Fluorometer, 100 - 240 VAC, Excitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nm

 
$2,226.42
   
Each
Minimum order quantity is 1
 

VWR® Fluorometer for quantitation of DNA, RNA, and Proteins. Compact with touch screen control panel. Compatible with VWR® quantitation kits and kits from leading suppliers.

  • Sensitive quantitation down to 0.1 pg/µl ds DNA

  • Large color touch screen for programming and operation
  • Two models for single or eight-sample processing
  • Built-in reagent calculators for sample preparation
  • Compact and portable (11×6" footprint)

Sample volume: 1 to 20 µl
Compatible sample tubes: 0.5 ml transparent thin wall PCR tube
Linearity range: Five orders of magnitude
Sensitivity: dsDNA: 0.01 ng/µl, ssDNA: 0.05 ng/µl, RNA: 0.25 ng/µl, microRNA: 0.05 ng/µl
Calibration: 2 or 3 points
Control panel: 5" color touch screen
Working environment: 15 to 30 ℃, <75% RH, indoor use

Certifications: Meets ASTM standards. EN-342.

Accessories information: Accessories not included.

 
  • Wavelength:
    Excitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nm
  • Description:
    VWR® Fluorometer, 100 - 240 VAC
  • Cat. No.:
    VWRI210-000157
  • Detector:
    Silicon photomultiplier
  • Scanning speed:
    ≤5 s/sample
  • W×D×H:
    10.8×24.1×5.7 cm (4.25×9.5×2.25")
  • Sample handling:
    1 (sample throughput)
  • Electrical:
    100 - 240 VAC
  • Light Source:
    LED
  • Weight:
    0.5 kg (1.1 lbs.)
Accessories

 

  •  
    Description
     
     
  • VWRI210-000159
    VWR® DS-DNA HS quant kit, 100 assays
       
    Each - $129.99

 

  •  
    Description
     
     
  • VWRI210-000160
    VWR® 1× ready-to-use DS-DNA hs quant kit, 100 assays
       
    Each - $135.20

 

  •  
    Description
     
     
  • VWRI210-000161
    VWR® DS-DNA BR quant kit, 100 assays
       
    Each - $135.19

 

  •  
    Description
     
     
  • VWRI210-000162
    VWR® 1× ready-to-use DS-DNA BR quant kit, 100 assays
       
    Each - $135.20

 

  •  
    Description
     
     
  • VWRI210-000163
    VWR® SS-DNA quant kit, 100 assays
       
    Each - $129.99

 

  •  
    Description
     
     
  • VWRI210-000164
    VWR® RNA HS quant kit, 100 assays
       
    Each - $129.99

 

  •  
    Description
     
     
  • VWRI210-000165
    VWR® RNA BR quant kit, 100 assays
       
    Each - $129.99

 

  •  
    Description
     
     
  • VWRI210-000166
    VWR® fluorometer tubes 0.5 ml, 250 per pack
       
    Pack of 250 - $72.80

 

  •  
    Description
     
     
  • VWRI210-000174
    Fluorometer tubes, 0.2 ml, pk/1000
       
    Each - $103.42

 

  •  
    Description
     
     
  • VWRI210-000175
    Fluorometer tube-strips, 0.2 ml, pk/120
       
    Each - $155.14

 

  •  
    Description
     
     
  • VWRI210-000177
    VWR® DS-DNA HS quant kit, 500 assays
       
    Each - $365.00

 

  •  
    Description
     
     
  • VWRI210-000178
    VWR® 1× DS-DNA HS quant kit, 500 assays
       
    Each - $385.00

 

  •  
    Description
     
     
  • VWRI210-000179
    VWR® DS-DNA BR quant kit, 500 assays
       
    Each - $365.00

 

  •  
    Description
     
     
  • VWRI210-000180
    VWR® 1× DS-DNA BR quant kit, 500 assays
       
    Each - $385.00

 

  •  
    Description
     
     
  • VWRI210-000181
    VWR® SS-DNA quant kit, 500 assays
       
    Each - $365.00

 

  •  
    Description
     
     
  • VWRI210-000182
    VWR® RNA HS quant kit, 500 assays
       
    Each - $365.00

 

  •  
    Description
     
     
  • VWRI210-000183
    VWR® RNA BR quant kit, 500 assays
       
    Each - $365.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.