Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Abgene™ Sample Processing and Storage Plates

Abgene™ Sample Processing and Storage Plates

Storage plate (Individual), 2200 µl, 96, V-conical, Natural, Cleanroom-manufactured, Square
Catalog # 77753-676
Supplier #:  AB-0932
Abgene™ Sample Processing and Storage Plates
Catalog # 77753-676
Abgene™ Sample Processing and Storage Plates
Abgene™ Sample Processing and Storage Plates

Storage plate (Individual), 2200 µl, 96, V-conical, Natural, Cleanroom-manufactured, Square

Price
$449.99
Log in for availability
Case of 50
Product Details & Documents

Abgene™ polypropylene MicroWell and DeepWell plates provide performance and value for sample storage, high-throughput screening, and molecular diagnostic assays. Abgene™ plates are made with high-quality, medical-grade, virgin polypropylene resin. These plates benefit from exceptional design and manufacturing advancements that ensure sample integrity and facilitate workflow efficiency.

  • Made of virgin, medical-grade polypropylene

  • hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
  • Low extractables and leachables
  • 48 to 384 well plate formats available
  • Temperature range: −80 to +121 °C
  • Volume range: 55 µl to 6 ml
  • ANSI/SLAS-compliant

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries, or storing samples for either intermediate or long-term use. Abgene™ microtiter plates are manufactured to exacting specifications in Class 100000 cleanroom ISO 9001 conditions using high-quality medical grade virgin USP <661.1> polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handlers for high-throughput workflows. These polypropylene storage plates are offered with spindle, pyramidal, U, and V bottom wells along with a wide range of volumes to accommodate most applications.

Packaging: Abgene™ Storage plates are available individually or bulk packed.

Specifications
  • No. of wells:
    96
  • Sterility:
    Cleanroom-manufactured
  • Color:
    Natural
  • Description:
    Storage plate (Individual)
  • Well shape:
    Square
  • Bottom style:
    V-conical
  • Total Volume:
    2200 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.