Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Abgene™ Sample Processing and Storage Plates

Storage plate (Individual), 800 µl, 96, V-conical, Natural, Cleanroom-manufactured, Round
Catalog # 77753-702
 :  AB-0765
Abgene™ Sample Processing and Storage Plates
Catalog # 77753-702
Abgene™ Sample Processing and Storage Plates
Abgene™ Sample Processing and Storage Plates

Storage plate (Individual), 800 µl, 96, V-conical, Natural, Cleanroom-manufactured, Round

Price
$313.10
Log in for availability
Case of 50
Product Details & Documents

Abgene™ polypropylene MicroWell and DeepWell plates provide performance and value for sample storage, high-throughput screening, and molecular diagnostic assays. Abgene™ plates are made with high-quality, medical-grade, virgin polypropylene resin. These plates benefit from exceptional design and manufacturing advancements that ensure sample integrity and facilitate workflow efficiency.

  • Made of virgin, medical-grade polypropylene

  • hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
  • Low extractables and leachables
  • 48 to 384 well plate formats available
  • Temperature range: −80 to +121 °C
  • Volume range: 55 µl to 6 ml
  • ANSI/SLAS-compliant

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries, or storing samples for either intermediate or long-term use. Abgene™ microtiter plates are manufactured to exacting specifications in Class 100000 cleanroom ISO 9001 conditions using high-quality medical grade virgin USP <661.1> polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handlers for high-throughput workflows. These polypropylene storage plates are offered with spindle, pyramidal, U, and V bottom wells along with a wide range of volumes to accommodate most applications.

 : Abgene™ Storage plates are available individually or bulk packed.

Specifications
  • No. of wells:
    96
  • Sterility:
    Cleanroom-manufactured
  • Color:
    Natural
  • Description:
    Storage plate (Individual)
  • Well shape:
    Round
  • Bottom style:
    V-conical
  • Total Volume:
    800 µl