Mouse Recombinant FGF-BASIC (from E. coli)
Vial, 1 mg
77752-028
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Pack type:Vial
- Size:1 mg
- Storage Conditions:–20 °C
- Molecular Weight:16.5 kDa
- Protease-free:N
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin Level:Low
- Purity:≥95%
- Carrier-Free:Y
- Source:E. coli
- Nuclease-free:N
- Biological Activity:Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
- Protein/Peptide Type:Recombinant
- Gene ID:P15655
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Protein/Peptide Name:FGF-BASIC
- Endotoxin-free:N
- Species:Mouse
- Protein Function:Cytokine
- Animal-Free:Y
- Tested Applications:Bioassay
- Grade:RUO
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Sequence:MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Cat. No.:77752-028
- Shipping Temperature:RT
- Protein Synonyms:FGF2|HBGF-2|Prostatropin
- Conjugation:Unconjugated



