Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse Recombinant SCF (from E. coli)

Mouse Recombinant SCF (from E. coli)

Vial, 500 µg
Catalog # 77751-992
Supplier:   VWR International
Mouse Recombinant SCF (from E. coli)
Catalog # 77751-992
Supplier:  VWR International
CAS Number:  
Mouse Recombinant SCF (from E. coli)
Mouse Recombinant SCF (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$1,508.33
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    18.4 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P20826
  • Formulation:
    20 mM Sodium phosphate, pH 6.5
  • Protein/Peptide Name:
    SCF (AF)
  • Endotoxin-free:
    N
  • Species:
    Mouse
  • Protein Function:
    Cytokine
  • Animal-Free:
    Y
  • Tested Applications:
    Bioassay
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
  • Cat. No.:
    77751-992
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    c-Kit Ligand|KL|Steel Factor|MGF
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.