Mouse Recombinant SCF (from E. coli)
Vial, 100 µg
Catalog # 77751-986
Supplier: VWR International
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
Specifications
- Pack type:Vial
- Size:100 µg
- Storage Conditions:–20 °C
- Molecular Weight:18.4 kDa
- Protease-free:N
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin Level:Low
- Purity:≥95%
- Carrier-Free:Y
- Source:E. coli
- Nuclease-free:N
- Biological Activity:Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
- Protein/Peptide Type:Recombinant
- Gene ID:P20826
- Formulation:20 mM Sodium phosphate, pH 6.5
- Protein/Peptide Name:SCF (AF)
- Endotoxin-free:N
- Species:Mouse
- Protein Function:Cytokine
- Animal-Free:Y
- Tested Applications:Bioassay
- Grade:RUO
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Sequence:MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
- Cat. No.:77751-986
- Shipping Temperature:RT
- Protein Synonyms:c-Kit Ligand|KL|Steel Factor|MGF
- Conjugation:Unconjugated
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



