Mouse Recombinant FGF-BASIC (from E. coli)
Vial, 100 µg
Catalog # 77752-024
Supplier: VWR International
Mouse Recombinant FGF-BASIC (from E. coli)
Catalog # 77752-024
Supplier: VWR International
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
Specifications
- Pack type:Vial
- Size:100 µg
- Storage Conditions:–20 °C
- Molecular Weight:16.5 kDa
- Protease-free:N
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin Level:Low
- Purity:≥95%
- Carrier-Free:Y
- Source:E. coli
- Nuclease-free:N
- Biological Activity:Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
- Protein/Peptide Type:Recombinant
- Gene ID:P15655
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Protein/Peptide Name:FGF-BASIC
- Endotoxin-free:N
- Species:Mouse
- Protein Function:Cytokine
- Animal-Free:Y
- Tested Applications:Bioassay
- Grade:RUO
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Sequence:MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Cat. No.:77752-024
- Shipping Temperature:RT
- Protein Synonyms:FGF2|HBGF-2|Prostatropin
- Conjugation:Unconjugated
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



