Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant GDF-15 (from E. coli)

Human Recombinant GDF-15 (from E. coli)

Vial, 20 µg
Catalog # 77750-694
Supplier:   VWR International
Human Recombinant GDF-15 (from E. coli)
Catalog # 77750-694
Supplier:  VWR International
CAS Number:  
Human Recombinant GDF-15 (from E. coli)
Human Recombinant GDF-15 (from E. coli)

Vial, 20 µg

CAS Number:  
Price
$163.33
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Growth differentiation factor 15 (GDF-15) is a member of the transforming growth factor beta (TGF-β) family and is made by the placenta and cardiovascular tissues. GDF-15 regulates inflammatory and apoptotic pathways during cellular stress and injury.

  • High quality
  • Low endotoxin
  • Affordable

GDF-15 is emerging as a biomarker of early heart disease, such that increased levels of circulating GDF-15 are associated with an increased risk of developing heart failure.

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    20 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    12.4/24.8 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    Q99988
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Protein/Peptide Name:
    GDF-15
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
  • Cat. No.:
    77750-694
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    MIC-1|Placental TGF-β|Prostate differentiation factor
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.