Human Recombinant FGF-BASIC 154 (from E. coli)
Vial, 50 µg
77750-420
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.
: For research use only.
- Pack type:Vial
- Size:50 µg
- Storage Conditions:–20 °C
- Molecular Weight:17.3 kDa
- Protease-free:N
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin Level:Low
- Purity:≥95%
- Carrier-Free:Y
- Source:E. coli
- Nuclease-free:N
- Biological Activity:Bioactivity assay: NR6R-3T3 Proliferation; ED50 ≤0.5 ng/ml; Bioactivity ≥2.0×10⁶ units/mg
- Protein/Peptide Type:Recombinant
- Gene ID:P09038
- Formulation:10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5
- Protein/Peptide Name:FGF-BASIC 154
- Endotoxin-free:N
- Species:Human
- Protein Function:Cytokine
- Animal-Free:N
- Tested Applications:Bioassay
- Grade:RUO
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Sequence:MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
- Cat. No.:77750-420
- Shipping Temperature:RT
- Protein Synonyms:FGF2|HBGF-2|Prostatropin
- Conjugation:Unconjugated



