Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant FGF-BASIC 154 (from E. coli)

Human Recombinant FGF-BASIC 154 (from E. coli)

Vial, 500 µg
Catalog # 77750-418
Supplier:   VWR International
Human Recombinant FGF-BASIC 154 (from E. coli)
Catalog # 77750-418
Supplier:  VWR International
CAS Number:  
Human Recombinant FGF-BASIC 154 (from E. coli)
Human Recombinant FGF-BASIC 154 (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$501.67
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state.

  • High quality
  • Low endotoxin
  • Affordable

Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    17.3 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity assay: NR6R-3T3 Proliferation; ED50 ≤0.5 ng/ml; Bioactivity ≥2.0×10⁶ units/mg
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P09038
  • Formulation:
    10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5
  • Protein/Peptide Name:
    FGF-BASIC 154
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Tested Applications:
    Bioassay
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • Cat. No.:
    77750-418
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    FGF2|HBGF-2|Prostatropin
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.