Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant GRO-G (from E. coli)

Human Recombinant GRO-G (from E. coli)

Vial, 500 µg
Catalog # 77750-684
Supplier:   VWR International
Human Recombinant GRO-G (from E. coli)
Catalog # 77750-684
Supplier:  VWR International
CAS Number:  
Human Recombinant GRO-G (from E. coli)
Human Recombinant GRO-G (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$1,765.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Growth regulated protein gamma (GRO-ɣ), also called CXCL3, acts through the chemokine receptor CXCR2 to promote monocyte migration and adhesion. GRO-ɣ also induces the migration of cerebellar granule neuron precursor cells.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Storage Conditions:
    −20 °C
  • Molecular Weight:
    7.9 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P19876
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Protein/Peptide Name:
    GRO-G
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN
  • Cat. No.:
    77750-684
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    MIP-2β|CXCL3|GRO3
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.