Human Recombinant GRO-G (from E. coli)
Vial, 1 mg
77750-682
Growth regulated protein gamma (GRO-ɣ), also called CXCL3, acts through the chemokine receptor CXCR2 to promote monocyte migration and adhesion. GRO-ɣ also induces the migration of cerebellar granule neuron precursor cells.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Pack type:Vial
- Size:1 mg
- Storage Conditions:−20 °C
- Molecular Weight:7.9 kDa
- Protease-free:N
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin Level:Low
- Purity:≥95%
- Carrier-Free:Y
- Source:E. coli
- Nuclease-free:N
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Protein/Peptide Type:Recombinant
- Gene ID:P19876
- Formulation:10 mM Sodium phosphate, pH 7.5
- Protein/Peptide Name:GRO-G
- Endotoxin-free:N
- Species:Human
- Protein Function:Cytokine
- Animal-Free:N
- Grade:RUO
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Sequence:ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN
- Cat. No.:77750-682
- Shipping Temperature:RT
- Protein Synonyms:MIP-2β|CXCL3|GRO3
- Conjugation:Unconjugated



