Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human Recombinant GRO-G (from E. coli)

Vial, 10 µg
Catalog # 77750-680
Supplier:   VWR International
Human Recombinant GRO-G (from E. coli)
Catalog # 77750-680
Supplier:  VWR International
CAS Number:  
Human Recombinant GRO-G (from E. coli)
Human Recombinant GRO-G (from E. coli)

Vial, 10 µg

CAS Number:  
Price
$163.33
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Growth regulated protein gamma (GRO-ɣ), also called CXCL3, acts through the chemokine receptor CXCR2 to promote monocyte migration and adhesion. GRO-ɣ also induces the migration of cerebellar granule neuron precursor cells.

  • High quality

  • Low endotoxin
  • Affordable

 : For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    10 µg
  • Storage Conditions:
    −20 °C
  • Molecular Weight:
    7.9 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P19876
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Protein/Peptide Name:
    GRO-G
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN
  • Cat. No.:
    77750-680
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    MIP-2β|CXCL3|GRO3
  • Conjugation:
    Unconjugated

Specifications