(+)-Biotin 4-Amidobenzoic Acid, Sodium Salt, TRC, LGC Standards
77693-947
New Product
:
- CAS No.:102418-74-6
- Description:(+)-Biotin 4-Amidobenzoic Acid, Sodium Salt
- Size:100 mg
- Formula:C₁₇H₂₀N₃NaO₄S
- Molecular Weight:385.41 g/mol
- Cat. No.:77693-947