Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TAG RAPID EAR PK200
Catalog # MSPP-RTA15
Supplier:
RAPID LAB INC MS
Expand image
TAG RAPID EAR PK200
Catalog # MSPP-RTA15
Supplier:
RAPID LAB INC MS
Supplier Number:
RTA15+