Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PH SINGLES POWDER BUFFER SOLUTION PK20

PH SINGLES POWDER BUFFER SOLUTION PK20

Catalog # 77674-496
Supplier:   CHEMetrics
Supplier #:  A-0175
PH SINGLES POWDER BUFFER SOLUTION PK20
Catalog # 77674-496
Supplier:  CHEMetrics
CAS Number:  
PH SINGLES POWDER BUFFER SOLUTION PK20
PH SINGLES POWDER BUFFER SOLUTION PK20
CAS Number:  
Price
$128.96
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.