Orders LinkContactUsLinkComponent
EVOM™ Auto Electrode Array for TEER
EVOM™ Auto Electrode Array for TEER
EVOM™ Auto 24C HTS electrode array for TEER
Catalog # 77664-276
Supplier #:  EVA-EL-03-02
EVOM™ Auto Electrode Array for TEER
Catalog # 77664-276
EVOM™ Auto Electrode Array for TEER
EVOM™ Auto Electrode Array for TEER
EVOM™ Auto 24C HTS electrode array for TEER
Price
$8,832.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EVOM™ Auto automates measurements of TEER in epithelial or endothelial monolayers cultured on high throughput screening (HTS) 24 and 96-well and plates utilizing our innovative EVOM technology, qualitatively measuring cell monolayer health and quantitatively measuring cell confluence by determining an increase or a plateau in tissue resistance.
  • Most reliable and convenient method of evaluating and monitoring the structural integrity of epithelial tissue culture in vitro
  • High-throughput 24 and 96-well plate system for labs with high volume
  • Unparalleled measurement accuracy with continuous data recording capability
  • Measure cellular activity label-free and real-time
  • 96 well plate electrode array is compatible with a wide variety of 96-well HTS plates: Corning, Millipore, and MatTek
  • 24C electrode array for Corning 24 HTS plates and 24M electrode array for 24 well Millipore 24 HTS plate
  • Easy switch and measurement in different plate types (24 or 96) by swapping electrode array and plate adapters using the same system
  • With web-based software, no operating system or security upgrades are required
Automated measurement of tissue resistance of transwell cell cultures provides the advantages of speed and precision, minimizes the chances of contamination, and ensures the rapid availability of measured resistance data. EVOM™ Auto produces a low AC current that omits any adverse physiological effects of the measurement technique on the samples and is specially designed for the non-destructive, high throughput screening of epithelial/ endothelial cellular physiology (e.g., confluence of cellular monolayers, barrier integrity of cell cultures, cytotoxicity).
Specifications
  • Description:
    EVOM™ Auto 24C HTS electrode array for TEER
Services
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 77664-274
    EVOM™ Auto 96 HTS electrode array for TEER-premium warranty 1 year
    Log in for availability
    Each - $3,079.69
  • 77664-278
    EVOM™ Auto 24C HTS electrode array for TEER-premium warranty 1 year
    Log in for availability
    Each - $3,079.69
  • 77664-282
    EVOM™ Auto 24M HTS electrode array for TEER-premium warranty 1 year
    Log in for availability
    Each - $3,079.69