Mini Iris Scissors
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Specifications
- Description:Mini iris scissor, sharp tips
- Length:8 cm (3.1")
- Material:Stainless steel
- Curvature:Curved