Orders LinkContactUsLinkComponent
Mini Dissecting Scissors
Mini Dissecting Scissors
Miniature dissecting scissors, 8.5 cm (3.3"), Stainless steel, Sharp, Curved
Catalog # 77663-340
Supplier #:  503666
Mini Dissecting Scissors
Catalog # 77663-340
Mini Dissecting Scissors
Miniature dissecting scissors, 8.5 cm (3.3"), Stainless steel, Sharp, Curved
Price
$79.20
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces.
They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 8.5 cm (3.3") long and have straight or curved blades with blunt or sharp tips and fine blades. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors.
They are designed for cutting and probing in tight spaces. These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Specifications
  • Description:
    Miniature dissecting scissors
  • Length:
    8.5 cm (3.3")
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Blade type:
    Sharp