Orders LinkContactUsLinkComponent
Operating Scissors
Operating Scissors
Operating scissors, 11.5 cm (4.5"), Stainless steel
Catalog # 77662-764
Supplier #:  501743
Operating Scissors
Catalog # 77662-764
Operating Scissors
Operating scissors, 11.5 cm (4.5"), Stainless steel
Price
$56.98
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled operating scissors are operating scissors are available with straight or curved blades commonly used in general surgery for cutting and dissecting tissues.
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
  • Autoclavable
That can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process.
Specifications
  • Description:
    Operating scissors
  • Length:
    11.5 cm (4.5")
  • Material:
    Stainless steel