Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Iris Scissors

Iris Scissors

Iris scissor, 45° angled, serrated tips, 11.5 cm (4.5"), Stainless steel, 2.2 cm
Catalog # 77663-158
Supplier #:  503225
Iris Scissors
Catalog # 77663-158
Iris Scissors

Iris scissor, 45° angled, serrated tips, 11.5 cm (4.5"), Stainless steel, 2.2 cm

Price
$120.13
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications.

  • Ring finger handle
  • Stainless steel

The tips are sharp, and they come in curved, straight or angled styles.

Specifications
  • Description:
    Iris scissor, 45° angled, serrated tips
  • Length:
    11.5 cm (4.5")
  • Material:
    Stainless steel
  • Blade length:
    2.2 cm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.