Orders LinkContactUsLinkComponent
Iris Scissors
Iris scissor, straight tips, 11.5 cm (4.5"), Stainless steel, 2.2 cm
Catalog # 77662-816
 :  501758
Iris Scissors
Catalog # 77662-816
Iris Scissors
Iris scissor, straight tips, 11.5 cm (4.5"), Stainless steel, 2.2 cm
Price
$73.94
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications.
  • Ring finger handle
  • Stainless steel
The tips are sharp, and they come in curved, straight or angled styles.
Specifications
  • Description:
    Iris scissor, straight tips
  • Length:
    11.5 cm (4.5")
  • Material:
    Stainless steel
  • Blade length:
    2.2 cm