Orders LinkContactUsLinkComponent
Metzenbaum Scissors
Metzenbaum scissors, straight tips, 17.5 cm (7"), German steel
  77662-514
 :  501254-G
Metzenbaum Scissors
  77662-514
Metzenbaum Scissors
Metzenbaum scissors, straight tips, 17.5 cm (7"), German steel
$53.00
Each
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.
  • Description:
    Metzenbaum scissors, straight tips
  • Length:
    17.5 cm (7")
  • Material:
    German steel