Orders LinkContactUsLinkComponent
Metzenbaum Scissors
Metzenbaum Scissors
Metzenbaum scissors, straight tips, 14.5 cm (5.75"), German steel
Catalog # 77662-506
Supplier #:  501252-G
Metzenbaum Scissors
Catalog # 77662-506
Metzenbaum Scissors
Metzenbaum scissors, straight tips, 14.5 cm (5.75"), German steel
Price
$50.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.
Specifications
  • Description:
    Metzenbaum scissors, straight tips
  • Length:
    14.5 cm (5.75")
  • Material:
    German steel